Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VXN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076475.1 | NP_001069943.1 | 3 - 4 | 502 | 74 | |
GenBank mRNA | BC119975.1 | - | 3 - 4 | 502 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |