Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GATA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076804.1 | NP_001070272.1 | 3 - 4 | 1338 | 61 | |
GenBank mRNA | BC123555.1 | - | 3 - 4 | 1338 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |