Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GATA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076804.1 | NP_001070272.1 | 4 - 5 | 1460 | 72 | |
GenBank mRNA | BC123555.1 | - | 4 - 5 | 1460 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |