Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC30A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076843.1 | NP_001070311.1 | 5 - 6 | 914 | 74 | |
GenBank mRNA | BC123455.1 | - | 5 - 6 | 914 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |