Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADGRL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076908.2 | NP_001070376.1 | 7 - 8 | 959 | 130 | |
GenBank mRNA | BC123530.1 | - | 7 - 8 | 960 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Mammalian Gene Collection Bos taurus Probe spans exons |