Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HID1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076924.1 | NP_001070392.1 | 10 - 11 | 1297 | 69 | |
GenBank mRNA | BC123459.1 | - | 10 - 11 | 1297 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |