Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MPPED2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076929.1 | NP_001070397.1 | 5 - 6 | 681 | 67 | |
GenBank mRNA | BC123804.1 | - | 5 - 6 | 681 | 67 | |
BC141996.1 | - | 6 - 7 | 949 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |