Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZMAT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076943.1 | NP_001070411.1 | 4 - 5 | 477 | 67 | |
GenBank mRNA | BC119856.1 | - | 4 - 5 | 477 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |