Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERPINA12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077517.1 | NP_001070985.1 | 2 - 3 | 849 | 84 | |
GenBank mRNA | BC123673.1 | - | 2 - 3 | 849 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |