Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPLX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077889.1 | NP_001071357.1 | 2 - 3 | 348 | 71 | |
GenBank mRNA | BC122691.1 | - | 2 - 3 | 348 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |