Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077933.2 | NP_001071401.1 | 4 - 5 | 680 | 70 | |
GenBank mRNA | BC120077.1 | - | 4 - 5 | 682 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Bos taurus Probe spans exons |