Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYOZ3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078039.1 | NP_001071507.1 | 6 - 7 | 812 | 130 | |
GenBank mRNA | BC123727.1 | - | 6 - 7 | 812 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |