Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC46A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079585.1 | NP_001073053.1 | 3 - 4 | 1184 | 89 | |
GenBank mRNA | BC122640.1 | - | 3 - 4 | 1184 | 89 | |
BT029834.1 | - | 3 - 4 | 1154 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |