Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAIM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079626.2 | NP_001073094.1 | 1 - 2 | 136 | 135 | |
GenBank mRNA | BC122668.1 | - | 1 - 2 | 137 | 135 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |