Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR83 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079650.2 | NP_001073118.1 | 7 - 8 | 738 | 76 | |
GenBank mRNA | BC120088.1 | - | 7 - 8 | 739 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |