Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIMMDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080235.2 | NP_001073704.1 | 3 - 4 | 660 | 96 | |
GenBank mRNA | BC122830.1 | - | 3 - 4 | 661 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |