Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COQ4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083672.2 | NP_001077141.1 | 3 - 4 | 340 | 73 | |
GenBank mRNA | BC122805.1 | - | 3 - 4 | 341 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |