Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173970.3 | NP_776395.1 | 5 - 6 | 638 | 129 | |
GenBank mRNA | AF503910.1 | - | 5 - 6 | 628 | 129 | |
BC120324.1 | - | 5 - 6 | 638 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |