Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GFAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174065.2 | NP_776490.2 | 1 - 2 | 459 | 80 | |
GenBank mRNA | BC119892.1 | - | 1 - 2 | 459 | 80 | |
Y08255.1 | - | 454 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |