Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGS16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174450.3 | NP_776875.1 | 3 - 4 | 313 | 78 | |
GenBank mRNA | AF011361.1 | - | 3 - 4 | 212 | 78 | |
BC123439.1 | - | 3 - 4 | 313 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |