Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CBY1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174739.4 | NP_777164.1 | 4 - 5 | 338 | 102 | |
GenBank mRNA | AF393210.1 | - | 5 - 6 | 566 | 102 | |
BC120055.1 | - | 4 - 5 | 339 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |