Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NFE2L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011678.2 | NP_001011678.2 | 1 - 2 | 137 | 113 | |
GenBank mRNA | BC102275.1 | - | 1 - 2 | 186 | 113 | |
BT030494.1 | - | 1 - 2 | 137 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |