Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035485.2 | NP_001030562.2 | 1 | 2447 | 152 | |
GenBank mRNA | BC123665.1 | - | 1 | 2447 | 152 | |
BT021198.1 | - | 1 | 2235 | 151 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Bos taurus Both primers and probe map within a single exon |