Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTDSP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045935.3 | NP_001039400.1 | 6 - 7 | 788 | 79 | |
GenBank mRNA | BC105531.1 | - | 6 - 7 | 650 | 79 | |
BC133616.1 | - | 6 - 7 | 789 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |