Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077020.1 | NP_001070488.1 | 8 - 9 | 850 | 87 | |
GenBank mRNA | BC105572.1 | - | 8 - 9 | 866 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |