Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EOGT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077882.1 | NP_001071350.1 | 14 - 15 | 1660 | 78 | |
GenBank mRNA | BC126625.1 | - | 14 - 15 | 1660 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |