Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
Gene Symbol: BCNT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077919.2 | NP_001071387.1 | 4 - 5 | 652 | 110 | |
GenBank mRNA | AB259013.1 | - | 4 - 5 | 640 | 110 | |
BC149202.1 | - | 4 - 5 | 625 | 110 | ||
EV657636.1 | - | 4 - 5 | 642 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |