Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077920.2 | NP_001071388.1 | 4 - 5 | 988 | 144 | |
GenBank mRNA | BC126548.1 | - | 4 - 5 | 989 | 144 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |