Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ENTPD8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077927.2 | NP_001071395.1 | 6 - 7 | 963 | 113 | |
GenBank mRNA | BC126637.1 | - | 6 - 7 | 964 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |