Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077965.2 | NP_001071433.1 | 1 - 2 | 88 | 63 | |
GenBank mRNA | BC126700.1 | - | 1 - 2 | 90 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |