Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCFD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078005.1 | NP_001071473.1 | 7 - 8 | 1936 | 98 | |
GenBank mRNA | BC126501.1 | - | 7 - 8 | 1936 | 98 | |
BC149466.1 | - | 4 - 5 | 1130 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |