Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLHL10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078033.2 | NP_001071501.1 | 4 - 5 | 1585 | 88 | |
GenBank mRNA | BC126695.1 | - | 4 - 5 | 1586 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |