Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BMERB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078077.1 | NP_001071545.1 | 4 - 5 | 480 | 113 | |
GenBank mRNA | BC126524.1 | - | 4 - 5 | 480 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |