Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIF3C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078155.1 | NP_001071623.1 | 7 - 8 | 2650 | 141 | |
GenBank mRNA | BC126505.1 | - | 7 - 8 | 2650 | 141 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |