Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TFAP2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080228.1 | NP_001073697.1 | 3 - 4 | 574 | 75 | |
GenBank mRNA | BC126845.1 | - | 3 - 4 | 574 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |