Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TFAP2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080228.1 | NP_001073697.1 | 4 - 5 | 801 | 124 | |
GenBank mRNA | BC126845.1 | - | 4 - 5 | 801 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |