Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080281.2 | NP_001073750.1 | 3 - 4 | 429 | 64 | |
GenBank mRNA | BC126650.1 | - | 3 - 4 | 430 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |