Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADRM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080282.1 | NP_001073751.1 | 4 - 5 | 515 | 75 | |
GenBank mRNA | BC149894.1 | - | 4 - 5 | 514 | 75 | |
BT029893.1 | - | 4 - 5 | 515 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |