Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C18H19orf47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080291.1 | NP_001073760.1 | 8 - 9 | 1097 | 111 | |
GenBank mRNA | BC126749.1 | - | 8 - 9 | 1097 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |