Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ODAM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080315.1 | NP_001073784.1 | 8 - 9 | 663 | 122 | |
GenBank mRNA | EF121760.1 | - | 8 - 9 | 663 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 Bos taurus Probe spans exons |