Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJC15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080332.2 | NP_001073801.1 | 3 - 4 | 246 | 114 | |
GenBank mRNA | BC126836.1 | - | 3 - 4 | 247 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |