Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIRPB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080911.1 | NP_001074380.1 | 4 - 5 | 1089 | 92 | |
GenBank mRNA | BT021649.1 | - | 4 - 5 | 1089 | 92 | |
BT026258.1 | - | 4 - 5 | 1088 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Bos taurus Probe spans exons |