Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LYSMD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081528.1 | NP_001074997.1 | 2 - 3 | 536 | 133 | |
GenBank mRNA | BC133419.1 | - | 2 - 3 | 536 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |