Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB40C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081545.1 | NP_001075014.1 | 6 - 7 | 736 | 112 | |
GenBank mRNA | BC133361.1 | - | 6 - 7 | 736 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |