Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC1A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081577.1 | NP_001075046.1 | 2 - 3 | 801 | 67 | |
GenBank mRNA | BC133295.1 | - | 2 - 3 | 801 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |