Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAGE4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081580.2 | NP_001075049.1 | 3 - 4 | 298 | 117 | |
GenBank mRNA | BC133479.1 | - | 3 - 4 | 299 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |