Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GABRD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081715.2 | NP_001075184.1 | 3 - 4 | 385 | 60 | |
GenBank mRNA | BC133557.1 | - | 3 - 4 | 386 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |