Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GABRD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081715.2 | NP_001075184.1 | 4 - 5 | 613 | 105 | |
GenBank mRNA | BC133557.1 | - | 4 - 5 | 614 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |