Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EFEMP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081717.2 | NP_001075186.1 | 10 - 11 | 1431 | 110 | |
GenBank mRNA | BC133561.1 | - | 10 - 11 | 1433 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |