Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC10A6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081738.1 | NP_001075207.1 | 766 | 82 | ||
GenBank mRNA | BC149199.1 | - | 3 - 4 | 825 | 82 | |
EF186076.1 | - | 766 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |